Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTY2D1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7143283
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9233 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9233 Supplier Bethyl Laboratories, Inc Supplier No. A304996AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Histone chaperone that stabilizes pre-existing histone tetramers and regulates replication-independent histone exchange on chromatin (PubMed:26109053). Required for normal chromatin refolding in the coding region of transcribed genes, and for the suppression of spurious transcription (PubMed:26109053). Binds DNA and histones and promotes nucleosome assembly (in vitro) (PubMed:23378026, PubMed:26109053). Facilitates formation of tetrameric histone complexes containing histone H3 and H4 (PubMed:26109053). Modulates RNA polymerase 1-mediated transcription. Binds DNA, with a preference for branched DNA species, such as Y-form DNA and Holliday junction DNA (PubMed:23378026).

Specifications

Antigen SPTY2D1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SPTY2D1
Gene Accession No. Q68D10
Gene Alias DKFZp686I068, FLJ39441, Protein KU002155, Spt2, SPT2 domain-containing protein 1, SPT2, Suppressor of Ty, domain containing 1, SPT2, Suppressor of Ty, domain containing 1 (S. cerevisiae)
Gene Symbols SPTY2D1
Host Species Rabbit
Immunogen Between 75 and 125
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144108
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.