Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Spunbond Polypropylene Shoe Cover

Catalog No. 19156242 Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-156-242 Blue Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-156-242 Supplier High Five™ Supplier No. EB1040000B

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Size Universal
Material Polypropylene
Color Blue
Closure Type Elastic Ankle
Model AS673
For Use With (Application) Ideal for protection against dirt, debris and light splashes
Resistance Abrasion
Product Type Shoe Covers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.