Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Heavy Duty Polypropylene Lab Coats
SDP

Catalog No. 19168685
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-685 3X-Large
19-168-681 Medium
19-168-682 Large
19-168-683 X-Large
19-168-684 2X-Large
19-168-686 4X-Large
19-168-687 5X-Large
19-168-680 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-168-685 Supplier Keystone Safety Supplier No. LC0NBENWHDV3XLNAVY
Add to Cart
Add to Cart
This item is not returnable. View return policy

Disposable lab coats are manufactured with strong, breathable, non-linting polypropylene fabric. Protects against dirt, grime, and dry particulates in non-hazardous environments.

  • Lightweight, breathable
  • Heavyweight disposable 40 gram polypropylene protective lab coats
  • No pockets, elastic wrists
  • Durable enough for multiple wearings but priced economically to allow disposal whenever necessary
  • Hook and loop fastening closure seals the front to reduce cross contamination
  • Class 10,000 (ISO 7) cleanroom compatible
  • Ideal for pharmaceutical, research, food service, dental and healthcare applications

Specifications

Product Type Controlled Environments Lab Coat
Size 3X-Large
Gender Unisex
Material Polypropylene
Color Navy
Cuff Style Elastic
Disposable Limited-use
Sterility Non-sterile
No. of Pockets None
Latex Free Yes
Model LCO-NBE-NW-HD-V
Certifications/Compliance Class 10,000, ISO 7
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Includes Thumb Loop
No. per Pack 30/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.