Learn More
Valutek™ Spunlace Non-woven Polyester Pre-Wetted Wiper, Cold knife cut
Description
- The pre-wetted format enhances the wipers absorbent capacity, effectiveness, and reduces airbone contamination from traditional solvent bottle dispensers, spray and spills
- 55% cellulose and 45% polyester blend with cold knife cut edge
- Size: 9 x 9 in.
- Contains no chemical binders
- Pre-wetted with specified IPA/DI Water concentrations to a 38% saturation level
- Available in both reusable pail packaging and refill pack packaging
- The pre-wet solution increases both cleaning efficiency and cleaning protocol consistency
- All Valutek™ pre-wetted wipers are assembled in a Class 10 cleanroom under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
- This product is recommended for use in a cleanroom Class 1-10 (ISO 3-4) critical environment
- It is designed for use in wet cleaning of critical surfaces and products
- Various IPA blend levels are ideal for removing different cleaning and disinfecting residues in regulated environments.
Specifications
Specifications
| Material | Polyester |
| Packaging Quantity | 75 /Pail, 2 Pails/Cs. |
| Saturation Solution | 90% IPA/10% DI H2O |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.