Learn More
Electron Microscopy Sciences Veco Square Center Reference 75 mesh Gold Grid

Supplier: Electron Microscopy Sciences 0075AU

Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Gold
Pitch: 333 µm
Hole: 283 µm
Bar: 50 µm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.