Learn More
Electron Microscopy Sciences Veco Square Center Reference 200 mesh Copper Grid

Supplier: Electron Microscopy Sciences 0200CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Grid: Copper
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.