Description
- Removes bacteria, particulates and macromolecules (such as pyrogens, nucleases and proteases) from ultrapure water.
- Easy replacement by simply twisting on and off dispenser.
- For use on the ultrapure dispensing module of Milli-Q™ SQ 2Series configurations, including Milli-Q™ SQ 200/200P and SQ 240C/240CV systems
Specifications
Specifications
| Filter Type | Final Filter |
| Type | Filter |