Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SQSTM1 Monoclonal Antibody (GT1478)
GREENER_CHOICE

Catalog No. PIMA527800 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527800 Supplier Invitrogen™ Supplier No. MA527800
  • $719.50
    $506.65 / Each of 1

    Save $212.85

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Positive Control: HepG2, A549, H1299, HCT116, Huh-7 (untreated), Huh-7 (3 μM Thapsigargin treatment for 12 hr), Huh-7 (3 μM Thapsigargin treatment for 24 hr), HepG2 (3 μM Thapsigargin treatment for 12 hr), HepG2 (3 μM Thapsigargin treatment for 24 hr), 2C4, MEF, MNT-1 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SQSTM1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT1478
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative; pH 7
Gene Sqstm1
Gene Accession No. Q13501, Q64337
Gene Alias A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3
Gene Symbols Sqstm1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human SQSTM1/P62. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18412, 8878
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.