Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Square HDPE Reagent Bottles with Wide Mouth Polypropylene Caps

Catalog No. 12141655 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
60 mL
175 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-655 60 mL
12-141-671 175 mL
12-141-656 250 mL
12-141-689 500 mL
12-141-672 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-141-655 Supplier Eisco Labs Supplier No. CH5318PK12
Only null left
Add to Cart
Add to Cart

The leak-proof design and wide mouth allow easy filling and pouring, with a -100°C to 120°C temperature range for versatile use.

  • Square reagent bottles - Wide mouth square HDPE reagent bottles with polypropylene caps, designed for easy handling and efficient storage in laboratory environments.
  • Leak-proof tested - Rigorously tested to ensure secure sealing under pressure, preventing spills during storage or transport.
  • Chemical resistance for versatile use - Made from HDPE, these bottles are ideal for storing a wide range of chemicals and reagents.
  • Laboratory and industrial applications - Perfect for chemical storage, liquid handling, and mixing in laboratories, research, and industrial settings.
  • Translucent and durable design - Lightweight, translucent construction provides visibility of contents, while the square shape optimizes storage space.
  • Designed for chemical storage and transport, these leak-proof, chemical-resistant bottles are ideal for laboratory and industrial applications. Non-sterile, -100°C to 120°C.

Specifications

Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Translucent
Material HDPE
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Product Type Reagent Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.