Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Square PET Media Bottles with Leakproof HDPE Caps

Catalog No. 12141643 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-643 125 mL
12-141-628 250 mL
12-141-662 500 mL
12-141-663 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-141-643 Supplier Eisco Labs Supplier No. CH5226PK24
Add to Cart
Add to Cart

Lab-grade, non-sterile bottles designed for secure storage of reagents, buffers, and biopharmaceutical solutions.

  • PET Media Bottles: Includes non-sterile PET bottles with secure HDPE caps, designed for laboratory reagents and buffer storage.
  • Leakproof Design: Semi-buttress threads and molded seal rings ensure reliable, spill-free storage and transport.
  • Efficient Storage Solution: Square-shaped ergonomic design for optimal storage space utilization and easy handling in labs.
  • Durable and Lightweight: Made of PET with heavy-duty walls, impact-resistant and suitable for routine lab applications.
  • Wide Temperature Range: Safe for use from -70°C to 60°C, supporting various scientific and biopharmaceutical processes.
  • Lightweight and durable.
  • Leakproof, tamper-evident caps ensure safe handling and transport for everyday lab use.

Specifications

Capacity (Metric) 125 mL
Capacity (English) 4.22 oz.
Color Translucent
Material PET
Sterility Non-sterile
Closure Included Yes
Disposable Single-use
Graduated No
Mouth Narrow
Shape Square
Product Type Media Bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.