Learn More
Description
- PETG Media Bottles: Includes non-sterile PETG bottles with secure HDPE caps, ideal for lab reagents, media, and buffer storage.
- Leakproof Design: Features semi-buttress threads and molded seal rings, providing reliable protection and preventing spills during storage or transport.
- Ergonomic Square Shape: Space-saving square design ensures efficient storage, with smooth surfaces for easy cleaning and residue-free use.
- Tamper-Evident Safety: Each bottle includes a heat-shrink band for added security, safeguarding stored materials from unauthorized access.
- Wide Temperature Range: Suitable for storage between -70°C and 60°C, supporting diverse lab and industrial applications with dependable performance.
- Designed for biopharmaceutical processing and reagent storage.
- Leak-proof, non-pyrogenic, and non-cytotoxic.
- Ideal for lab and industrial applications requiring secure, reliable storage.
- Leakproof with tamper-evident caps, they provide reliable handling, efficient storage, and easy identification in lab environments.
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Capacity (English) | 1 oz. |
| Color | Translucent |
| Material | PETG |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Disposable | Single-use |
| Graduated | No |
| Mouth | Narrow |
| Shape | Square |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.