Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Square Polypropylene Reagent Bottles with Wide Mouth Polypropylene Caps

Catalog No. 12141636 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
60 mL
175 mL
250 mL
500 mL
1000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-636 60 mL
12-141-634 175 mL
12-141-686 250 mL
12-141-687 500 mL
12-141-670 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-141-636 Supplier Eisco Labs Supplier No. CH5311PK12
Only null left
Add to Cart
Add to Cart

Translucent, chemical-resistant bottles that are leak-proof and autoclavable, capable of withstanding temperatures from 0 to 135 °CC.

  • Square bottles - Square polypropylene reagent bottles with wide mouths and polypropylene caps, offering optimal storage for lab reagents and chemicals.
  • Leak-proof design - Engineered with a secure seal to prevent leaks during storage or transport rigorously tested.
  • Autoclavable and chemical-resistant - Made from polypropylene, these bottles can be sterilized up to 135°C and resist a variety of laboratory chemicals.
  • Versatile laboratory use - Ideal for storing, mixing, and transporting liquids in research, medical, and industrial settings. Square shape enhances storage efficiency.
  • Translucent with easy handling - Translucent design allows for quick content visibility, and lightweight construction with wide necks ensures easy filling and pouring.

Specifications

Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Translucent
Material Polypropylene
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Product Type Reagent Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.