Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squaric Acid Dibutyl Ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2892-62-8
C12H18O4
226.27
MFCD00037150
XBRWELTXMQSEIN-UHFFFAOYSA-N
dibutyl squarate,3,4-dibutoxy-3-cyclobutene-1,2-dione,squaric acid dibutyl ester,sadbe,squaric acid dibutylester,3,4-di-n-butoxy-3-cyclobutene-1,2-dione,unii-4rto57vg65,1,2-dibutyl squarate,3-cyclobutene-1,2-dione, 3,4-dibutoxy,acmc-209h5p
65108
CHEBI:53612
3,4-dibutoxycyclobut-3-ene-1,2-dione
CCCCOC1=C(C(=O)C1=O)OCCCC
2892-62-8
C12H18O4
226.27
MFCD00037150
XBRWELTXMQSEIN-UHFFFAOYSA-N
dibutyl squarate,3,4-dibutoxy-3-cyclobutene-1,2-dione,squaric acid dibutyl ester,sadbe,squaric acid dibutylester,3,4-di-n-butoxy-3-cyclobutene-1,2-dione,unii-4rto57vg65,1,2-dibutyl squarate,3-cyclobutene-1,2-dione, 3,4-dibutoxy,acmc-209h5p
65108
CHEBI:53612
3,4-dibutoxycyclobut-3-ene-1,2-dione
CCCCOC1=C(C(=O)C1=O)OCCCC
Infrared Spectrum Authentic
Quantity 25g
Color Colorless to Yellow
Specific Gravity 1.05
Flash Point >110°C
Packaging Glass bottle
Density 1.0500g/mL
Refractive Index 1.4930 to 1.4950
Boiling Point 121.0°C to 122.0°C (0.2 mmHg)
Assay Percent Range 97%
Merck Index 15, 8897
Solubility Information Solubility in water: insoluble.
Formula Weight 226.27
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Squaric acid dibutyl ester
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning