Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3000 Retractable Tool Lanyard with Dual Stainless Steel Carabiners

Catalog No. 19820313 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-313 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-313 Supplier Ergodyne™ Supplier No. 19300
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Squids™ 3000 Retractable Tool Lanyard with Dual Stainless Steel Carabiners is designed to mount to the users belt or a separate anchor point.

  • Retractable design limits snags, tangles and tripping hazardsModular connecting buckle to quickly exchange tools when used together with the Squids™ 3025 Stainless Steel Carabiner Accessory 3-Pack
  • Swiveling ring for 360-degree rotation works well with rotating tools to reduce tangles
  • Self-setting pawls set desired lanyard length to take the tension out
  • ABS thermoplastic housing protects the internal components and Technora cord for ultimate strength and durability

Specifications

Color Black
Load Bearing Capacity (Metric) 0.5 kg
Size Standard
Load Bearing Capacity (English) 1 lb.
Length (Metric) 122 cm
Length (English) 48 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.