Learn More
Ergodyne™ Squids™ 3026 Accessory Pack Retractables
Catalog No. 19820318 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Black
Catalog No.
19-820-318 Color: Black
1 options
Catalog No. 19-820-318 Supplier Ergodyne™ Supplier No. 19326
Squids™ 3026 Accessory Pack Retractables includes 3 tails with choking loop attachments designed to mount to the Squids™ 3001 Retractable Tool Lanyard.
  • Choking loop made with an energy-absorbing cord and tubular nylon webbing connects to tools
  • Modular connecting buckle to quickly exchange tools
Specifications
Color Black
Certifications/Compliance ANSI/ISEA 121-2018
Size Standard
Load Bearing Capacity (Metric) 0.9 kg
Load Bearing Capacity (English) 2 lb.