Learn More
Ergodyne™ Squids™ 3109F(x) Double-Locking Single Swivel Carabiner Tool Lanyard
Shop All Ergodyne Products
Description
- Approved to the ANSI/ISEA 121-2018 standard
- Shock absorbing properties reduce the dynamic force on the body or other anchor point
- Durable 1680 denier polyester elastic blended webbing extends from 38 to 48 in. (96 to 122 cm)
- Low profile lightweight design reduces snag hazards and added weight
- Anodized aluminum alloy swiveling carabiner with double action locking gate and inherent captive eye
- Ergonomic carabiner is designed for easily gripping and handling with gloves
- Treaded loop end that grips the tool when attached
- High strength 3-ply nylon stitching secures connecting ends
- Single length construction eliminates weak points seen with multi-component lanyards
Specifications
Specifications
| Color | Orange, Gray |
| Size | Standard |
| Certifications/Compliance | ANSI/ISEA 121-2018 standard |