Learn More
Ergodyne™ Squids™ 3119F(x) Double-Locking Dual Swivel Carabiner Tool Lanyard
Catalog No. 19820366 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Orange, Gray
Catalog No.
19-820-366 Color: Orange, Gray
1 options
Catalog No. 19-820-366 Supplier Ergodyne™ Supplier No. 19829
Squids™ 3119F(x) Double-Locking Dual Swivel Carabiner Tool Lanyard is designed to prevent falling objects.
  • Shock absorbing properties reduce the dynamic force on the body or other anchor point
  • Durable 1680 denier polyester elastic blended webbing extends from 38 to 48 in. (96 to 122 cm)
  • Low profile lightweight design reduces snag hazards and added weight
  • Anodized aluminum alloy swiveling carabiners with double action locking gate and inherent captive eye
  • Ergonomic carabiners are designed for easily gripping and handling with gloves
  • High strength 3-ply nylon stitching secures connecting ends
  • Single length construction eliminates weak points seen with multi-component lanyards
Specifications
Color Orange, Gray
Size Standard
Certifications/Compliance ANSI/ISEA 121-2018 standard