Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ Squids™ 3157 Coil Tool Lanyard with Buckle
Squids™ 3157 Coil Tool Lanyard with Buckle is compact and used to tether small hand tools, which prevents falling objects when working at heights.
$51.20
Specifications
Certifications/Compliance | Approved to the ANSI/ISEA 121-2018 standard |
---|---|
Length (English) | 12 to 48 in. |
Length (Metric) | 30 to 122 cm |
Load Bearing Capacity (English) | 2 lb. |
Load Bearing Capacity (Metric) | 0.9 kg |
Catalog Number | Mfr. No. | dimColor | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimColor | Price | Quantity & Availability | |||||
19-910-452
|
Ergodyne™
19156 |
Each for $51.20
|
|
||||||
19-910-453
|
Ergodyne™
19157 |
Each for $51.20
|
|
||||||
Description
- Approved to the ANSI/ISEA 121-2018 Standard
- Low-profile durable plastic coil to withstand even the toughest conditions
- Easily attaches to hard hats and other small hand tools for safekeeping
- Coiled lanyard design prevents snag and tangle hazards
- Double lock buckle attaches to harness
- Tethering loop to securely connect to an anchor or tool attachment
- Certified to a 2:1 safety factor
Specifications
Approved to the ANSI/ISEA 121-2018 standard | |
30 to 122 cm | |
0.9 kg |
12 to 48 in. | |
2 lb. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title