Learn More
Ergodyne™ Squids™ 3400 Metal Detectable Grabber
Catalog No. 19820308 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Blue
Catalog No.
19-820-308 Color: Blue
1 options
Catalog No. 19-820-308 Supplier Ergodyne™ Supplier No. 19132
Squids™ 3400 Metal Detectable Grabber keeps gloves, rags, keys and other pieces of equipment safe and secure.
  • Grabber clip mounts to d-rings, clothing, etc.
  • Metal detectable material alerts quality control
  • Entire piece impregnated with metallic powder
  • Grabbers disconnect and can be used independently
  • Deep blue color signals metal detectability
  • Breakaway feature
Specifications
Color Blue
Material Metal