Learn More
Ergodyne™ Squids™ 3740 Hand Tool Attachment Trap Slips™
Catalog No. 19820339 Shop All Ergodyne Products
Change view
Click to view available options
Includes:
4/Pk.
60/Pk.
Size:
Large
Medium
Small
Variety
X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
19-820-339 4/Pk. Small
19-820-340 4/Pk. Medium
19-820-341 4/Pk. Large
19-820-342 4/Pk. X-Large
19-820-343 4/Pk. Variety
19-820-378 60/Pk. Small
19-820-379 60/Pk. Medium
19-820-380 60/Pk. Large
19-820-381 60/Pk. X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-820-339 Supplier Ergodyne™ Supplier No. 19740
Add to Cart
Add to Cart
Squids™ Slips™ are retrofit tool attachments for small hand tools, which help reduce the risk of dropped objects and increase workplace safety.
  • Approved to the ANSI/ISEA 121-2018 standard
  • One-piece design easily slides onto small hand tools without requiring additional tools to install
  • Swiveling exterior ring spins 360 degrees, allowing screwdrivers and other rotating tools to be used without issue
  • ABS outer ring provides strength and durability in all environments
  • Flexible inner core grips tool after application
  • Intended for singled use
Specifications
Color Black, Orange
Includes 4/Pk.
Load Bearing Capacity (Metric) 0.45 kg
Size Small
Quantity 4/Pk.
Load Bearing Capacity (English) 1 lb.
Certifications/Compliance ANSI/ISEA 121-2018 standard