Learn More
Ergodyne™ Squids™ 3770 Tape Measure Holder and Belt Clip
Catalog No. 19820352 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Orange
Catalog No.
19-820-352 Color: Orange
1 options
Catalog No. 19-820-352 Supplier Ergodyne™ Supplier No. 19770
Squids™ 3770 Tape Measure Holder and Belt Clip is designed with a zinc-plated cast alloy D-Ring connection point.
  • 1200D and 600D polyester main material for long life and durability
  • Hook and loop straps secure tape measure inside trap for safe and easy tethering
  • Includes strong zinc-plated cast alloy D-Ring for tethering
  • Stainless steel belt clip securely attaches to a tool belt
  • Fits tape measures from 16 to 25 ft. (5 to 8 m)
Specifications
Color Orange
Material Polyester (1200D), Polyester (600D)
Size Large
Certifications/Compliance ANSI/ISEA 121-2018 standard