Learn More
Ergodyne™ Squids™ 3775 Bottle Holder and Trap
Catalog No. 19820353 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Black
Catalog No.
19-820-353 Color: Black
1 options
Catalog No. 19-820-353 Supplier Ergodyne™ Supplier No. 19775
Squids™ 3775 Bottle Holder and Trap makes it easy to carry and secure your water bottle, spray bottle, or other canisters.
  • Neoprene material is abrasion-resistant and stretches to fit a variety of bottles/cans
  • Heavy gauge nylon stitching for extra reinforcement
  • Convenient stainless steel belt clip attaches to tool belts and fall protection harnesses
  • Zinc-plated D-ring for tool lanyard attachment
  • Adjustable buckle closure to securely hold the bottle/can in the holder
  • Fits standard 16.9 oz. (0.5 L) water bottles
Specifications
Color Black
Certifications/Compliance ANSI/ISEA 121-2018 standard
Load Bearing Capacity (Metric) 0.9 kg
Load Bearing Capacity (English) 2 lb.