Learn More
Ergodyne™ Squids™ 3790 Tool Shackle
Catalog No. 19820356 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-356 Small
19-820-357 Medium
19-820-358 Large
19-820-359 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-820-356 Supplier Ergodyne™ Supplier No. 19792
Only null left
Add to Cart
Add to Cart
Squids™ 3790 Tool Shackles connect to waisted/necked tools or tools with captive holes, creating clean and secure attachment points for tethering.
  • Approved to the ANSI/ISEA 121-2018 standard
  • 304 stainless steel material
  • One step retrofit attachment for tethering point
  • Connects around midsection of a waisted or necked tool
  • Secure by adding a drop of adhesive and tightening
Specifications
Load Bearing Capacity (Metric) 6.8 kg
Includes 2/Pk.
Load Bearing Capacity (English) 15 lb.
Size Small
Certifications/Compliance Approved to the ANSI/ISEA 121-2018 standard