Learn More
Ergodyne™ Squids™ 3790 Tool Shackle
Catalog No. 19820359 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-359 X-Large
19-820-356 Small
19-820-357 Medium
19-820-358 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-820-359 Supplier Ergodyne™ Supplier No. 19795
This item is not returnable. View return policy
Squids™ 3790 Tool Shackles connect to waisted/necked tools or tools with captive holes, creating clean and secure attachment points for tethering.
  • Approved to the ANSI/ISEA 121-2018 standard
  • 304 stainless steel material
  • One step retrofit attachment for tethering point
  • Connects around midsection of a waisted or necked tool
  • Secure by adding a drop of adhesive and tightening
Specifications
Color Gray
Material Stainless Steel
Size X-Large
Load Bearing Capacity (English) 15 lb.
Includes 2/Pk.
Certifications/Compliance Approved to the ANSI/ISEA 121-2018 standard
Load Bearing Capacity (Metric) 6.8 kg