Learn More
Ergodyne™ Squids™ 3797 Power Tool Bracket with Grinder Tool Trap™
Catalog No. 19820361 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Gray
Catalog No.
19-820-361 Color: Gray
1 options
Catalog No. 19-820-361 Supplier Ergodyne™ Supplier No. 19797
Squids™ 3797 Power Tool Bracket with Grinder Tool Trap™ creates a unique tool attachment point to tether corded and cordless grinder power tools.
  • Stainless steel bracket with built-in connection point for tool tethering is corrosion-resistant to withstand weathering
  • Tool bracket design fastens to open screw port on grinders and directionally pushes the lanyard away from the grinder's wheel
  • Perfect for corded or cordless angle grinders and similar power tools
  • Ambidextrous design can be flipped to either side of the tool attaching to open screw ports meant for grinder handles
  • Fasteners include stainless steel M8 and M10 hex head bolts
  • Maximum tethering capacity of 8 lb. (3.63 kg)
Specifications
Color Gray
Material Stainless Steel
Certifications/Compliance ANSI/ISEA 121-2018 standard
Includes 1 bracket connection point, 2 fasteners and locking washers, and 1 product label