Learn More
Ergodyne™ Squids™ 3798 Power Tool Bracket with Pneumatic Tool Trap™
Catalog No. 19820362 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Gray
Catalog No.
19-820-362 Color: Gray
1 options
Catalog No. 19-820-362 Supplier Ergodyne™ Supplier No. 19798
Squids™ 3798 Power Tool Bracket with Pneumatic Tool Trap™ creates a unique tool attachment point to tether pneumatic power tools.
  • Stainless steel bracket with built-in connection point for tool tethering is corrosion-resistant to withstand weathering
  • Tool bracket design fastens onto pneumatic/air powered tools using the tools air hose coupler
  • Ideal for a variety of pneumatic tools using an air hose like angle grinders, paddle sanders, handheld buffers, nailers, staplers, and rivet guns
  • Fits couplers ranging in size from 0.25 in. (6.3 mm) to 0.375 in. (9.5 mm)
  • Maximum tethering capacity of 5 lb. (2.26 kg)
Specifications
Color Gray
Material Stainless Steel
Includes 1 bracket connection point and 1 product label
Certifications/Compliance ANSI/ISEA 121-2018 standard