Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3191 Power Tool Tethering Kit - 10lbs/4.5kg

Catalog No. 19820323 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
power tool kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-323 power tool kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-820-323 Supplier Ergodyne™ Supplier No. 19661
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tool Tethering Kit allows users to tether power tools, making it easy to use tools on the job while preventing dropped objects when working at-heights.

  • Product solutions for tethering cordless power drills and cordless impact drivers
  • Straightforward installation
  • Lack of tool lanyards can create a hazardous environment while working at heights
  • No tape and tool tails required
  • Open up the hook and loop straps of the power tool trap and place the battery end of the power tool into the power tool trap, with the back of the battery near the D-ring of the trap. Tighten the hook-and-loop straps on the power tool trap to secure the power tool inside. Attach the 3100F(x) Tool Lanyard to complete your tethering system

Specifications

Load Bearing Capacity (English) 10 lb.
Certifications/Compliance ANSI/ISEA 121-2018
Includes Squids™ 3100F(x) Single Carabiner Tool Lanyard and a Squids™ 3780L Power Tool Trap
Product Type Squids™ 3780 Power Tool Trap™
Load Bearing Capacity (Metric) 4.5 kg
Size One tool
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.