Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3190 Tape Measure Tethering Kit - 2lbs/0.9kg

Catalog No. 19820322 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
2 lb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-820-322 2 lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-820-322 Supplier Ergodyne™ Supplier No. 19660
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tool Tethering Kit allows users to tether tape measures making it easy to use tools on the job while preventing dropped objects when working at-heights.

  • Straightforward installation
  • Lack of tool lanyards can create a hazardous environment while working at heights
  • No tape and tool tails required
  • Open up the hook-and-loop straps. Place the tape measure inside the trap, with the back of the tape measure facing the D-ring on the back of the trap. Adjust and tighten the hook-and-loop straps accordingly. Attach the Coil Tool Lanyard to the D-ring to complete the tethering system

Specifications

Load Bearing Capacity (English) 2 lb.
Load Bearing Capacity (Metric) 0.9 kg
Product Type Squids™ 3770 Tape Measure Trap
Certifications/Compliance ANSI/ISEA 121-2018
Size Tether one tape measure
Includes Squids™ 3130S Coil Lanyard and a Squids™ 3770 Tape Measure Trap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.