Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ Tool Tethering Kit

Catalog No. 19820319 Shop All Ergodyne Products
Change view
Click to view available options
Load Bearing Capacity (English):
Up to 3 x 10 lb. tools
Up to 4 x 5 lb. tools
Up to 6 x 2 lb. tools
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (English)
19-820-319 Up to 6 x 2 lb. tools
19-820-320 Up to 4 x 5 lb. tools
19-820-321 Up to 3 x 10 lb. tools
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-820-319 Supplier Ergodyne™ Supplier No. 19650
Add to Cart
Add to Cart

Tool Tethering Kit allows users to tether tools, making it easy to use tools on the job while preventing dropped objects when working at-heights.

  • Straightforward installation
  • Lack of connection points for tool lanyards can create a hazardous environment while working at heights
  • Measure and cut tape to the specified length listed in instructions. Lay the tool tail on the tool being tethered and wrap the tape once around itself beginning below one of the web catch points. Continue wrapping and stretching the tape firmly. Firmly push down on the end and attach the Squids™ Tool Lanyard to complete the tethering system.

Specifications

Load Bearing Capacity (English) Up to 6 x 2 lb. tools
Includes Squids™ 3130S Coil Lanyard, 12 ft / 3.7 m roll of Squids™ 3755 Self-Adhering Tape Trap, and 6 Squids™ 3700 Web Tool Tails
Load Bearing Capacity (Metric) Up to 6 0.9 kg tools
For Use With (Equipment) Small hand tools including: pliers, claw hammers, sledge hammers, cat’s paws, utility knives, jab/drywall saws, caulking knives, nips and trowels
Certifications/Compliance ANSI/ISEA 121-2018
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.