Learn More
Enzo Life Sciences SR-12813 (50mg). CAS: 126411-39-0
SDP
Supplier:  Enzo Life Sciences BMLGR2420050
Encompass_Preferred
Potent and selective PXR agonist. (EC50=200 nM for human PXR). A weak activator of rat and mouse PXR. Inhibits cholesterol biosynthesis in a number of species. Alternative name: Tetraethyl 2-(3,5-Di-t-butyl-4-hydroxyphenyl)ethenyl-1,1-bisphosphonate. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0946
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.