Learn More
Selleck Chemical LLC SR-18292 10mg 2095432-55-4
SDP
Supplier:  Selleck Chemical LLC S852810MG
Encompass_Preferred
SR-18292 inhibits PGC-1? gluconeogenic activity and reduces co-activation of HNF4? by modulating the interaction between GCN5 and PGC-1?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8150
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.