Learn More
Selleck Chemical LLC SR-3029
Supplier: Selleck Chemical LLC S65445MG
SR-3029 is a potent and selective casein kinase 1 /1 (CK1 / ) inhibitor with IC50 values of 44 and 260 nM for CK1 and CK1 respectively