Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 48692
GREENER_CHOICE

Catalog No. 372125RD
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-212-5RD 25 mg
37-215 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-212-5RD Supplier Tocris Bioscience™ Supplier No. 3721/25
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective non-peptide NTS1 antagonist

SR 48692 is a neurotensin antagonist; selective for NTS1 over NTS2 (apparent affinity, Ke, is 36 nM for NTS1). Competitively inhibits binding of [125I]-neurotensin to HT29 and N1E115 cell membranes (IC50 values are 15.3 and 20.4 nM respectively). Orally bioavailable.

Specifications

Chemical Name or Material 2-[[[1-(7-Chloro-4-quinolinyl)-5-(2,6-dimethoxyphenyl)-1H-pyrazol-3-yl]carbonyl]amino]-tricyclo[3.3.1.13,7]decane-2-carboxylic acid
CAS 146362-70-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C32H31ClN4O5
Formula Weight Observed MW: 587.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.