Learn More
Stoelting™ SR-LAB™ Startle Response System
SDP
Catalog No. 10001055 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
SR-LAB Large Animal Enclosure
SR-LAB Medium Animal Enclosure
SR-LAB S Animal Encl w/Ultrasensor
SR-LAB S/M Animal Encl w/ULTsensor
SR-LAB System for Desktop Computer
SR-LAB System, Additional Unit
SR-LAB X-Large Animal Enclosure
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-001-055 SR-LAB System, Additional Unit
10-001-054 SR-LAB System for Desktop Computer
10-001-056 SR-LAB S Animal Encl w/Ultrasensor
10-001-057 SR-LAB S/M Animal Encl w/ULTsensor
10-001-058 SR-LAB Medium Animal Enclosure
10-001-059 SR-LAB Large Animal Enclosure
10-001-060 SR-LAB X-Large Animal Enclosure
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-001-055 Supplier Stoelting™ Supplier No. 59802
Only null left
Add to Cart
Add to Cart
Request a Quote
SR-LAB™ Startle Response System features a design that ensures accurate results for all paradigms and is ideal for startle reflex testing
  • Easy to use with straightforward assembly and menu-driven software
  • Expands to include 16 test stations
  • Use in all startle paradigms including startle habituation, pre-pulse, and cross modal inhibition
  • Standardized test cabinets
  • Basic Startle Reflex hardware and Microsoft Windows based software give users complete control over startle paradigms. Virtually any combination of tones, noise bursts, lights, air puffs, background noise and foot shock options are controllable.
  • Supports up to 16 test stations.
  • Test cabinets allow full and unobtrusive observation of the test animals.
  • Data collected are completely secure and recorded in a non-editable form
SR-LAB Components basic system components include:
  • Test cabinet in composite ABS plastic
  • Animal enclosure in choice of standard sizes
  • Power Supply
  • Input and Output cables
  • Software and User Manual
  • Software utilities for data analysis
  • Animal Enclosures
  • Animal enclosures are designed to locate the subject without using restraint. The enclosure monitors movements with an ultra-stable, hermetically sealed motion sensor using new USB 16 bit resolution board (a range from zero to over 10,000 millivolts. A 16 bit board diveds 10,000 mV by 65536 discrete numbers. Thus, 10,000/65536 = 0.15 MV and now you get a new data point with just a 0.15 mV change) for measuring startle responses. All enclosures are adjustable in length and made of transparent acrylic for easy cleaning and convenient observation.
Software Features
The SR-Lab Software helps users plan, execute and manage data in startle reflex testing sessions. Users can define trial definitions and session definitions with a user-friendly interface. Test progress can be displayed with summary numeric data and response waveforms. After the session has ended, the complete waveform can be reviewed, and data may be refined via settable parameters including Baseline, Onset Window Start, End Analysis, and Onset criterion. The software stores all data in a database for easy exporting to Microsoft Excel and/or statistical software programs.
Specifications
For Use With (Application) Neuroscience
Model SR-LAB System, Additional Unit