Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SR140 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541970 Supplier Invitrogen™ Supplier No. PA541970
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NLSRPLLENK LKAFSIGKMS TAKRTLSKKE QEELKKKEDE KAAAEIYEEF Sequence homology: Cow: 100%; Dog: 100%; Goat: 80%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 86%.

U2SURP (U2 SnRNP Associated SURP Domain Containing) is a Protein Coding gene. Diseases associated with U2SURP include Retinitis Pigmentosa 67. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and RNA binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR140
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene U2SURP
Gene Accession No. O15042
Gene Alias 140 kDa Ser/Arg-rich domain protein; 2610101N10Rik; AU023006; fSAPa; functional spliceosome-associated protein a; KIAA0332; mp:zf637-1-000918; RGD1307882; Ser/Arg-rich domain protein, 140 kDa; si:dkey-181m9.9; SR140; U2 associated SR140 protein; U2 snRNP associated SURP domain containing; U2 snRNP-associated SURP domain containing; U2 snRNP-associated SURP motif-containing protein; U2-associated protein SR140; u2-associated protein SR140-like protein; U2-associated SR140 protein; U2SURP; wu:fa03e02; wu:fb38b01; wu:fc75d03
Gene Symbols U2SURP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SR140 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23350
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.