Learn More
TARGETMOL CHEMICALS INC SR3335 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5160
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR3335 (ML 176) is a selective inverse agonist of ROR(alpha) competitively inhibiting the binding of 25-hydroxycholesterol to the ligand binding domain (Ki 220 nM). purity: 99%
Catalog No. 50-257-6703
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.