Learn More
Eurofins Discoverx Src (1-530), active, 10 µg. (14-746)
Supplier:  Eurofins Discoverx 14746
Encompass
N-terminal 6xHis-tagged recombinant, human Src amino acids 1-530. The mutant form of Src (Src, 1-530) has been identified in cases of advanced colon cancer. This truncating mutation generates an open active form of Src that has been found to be transforming, tumorigenic and metastasis promoting. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9279
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.