Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Src I1
GREENER_CHOICESDP

Catalog No. 364210R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-421-0R 10 mg
36-425-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-421-0R Supplier Tocris Bioscience™ Supplier No. 3642/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dual site Src kinase inhibitor

Src I1 is a potent, competitive dual site (ATP- and peptide-binding) Src kinase inhibitor (IC50 values are 44 and 88 nM for Src and Lck respectively). Inhibits VEGFR2 and c-fms at higher concentrations (IC50 values are 0.32 and 30 μM respectively). Can be used in parallel with PP 1 (Cat. No. 1397) and PP 2 (Cat. No. 1407) to inhibit Src family kinases.

Specifications

Quantity 10 mg
Product Type Src I1
Purity >99%
Molecular Weight (g/mol) 373.4
Inhibitors Src I1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.