Learn More
Description
PA1-840 detects steroid receptor coactivator-1 (SRC-1) from human, mouse, and japaneese quail tissues and cells. PA1-840 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry procedures. By Western blot, this antibody detects an ~165 kDa protein representing SRC-1 and a smaller, unknown protein at ~40 kDa from HeLa cell lysate. Immunocytochemical staining of SRC-1 in HeLa cells with PA1-840 results in nuclear staining. The PA1-840 immunogen is a synthetic peptide corresponding to residues S(8) S D P A N P D S H K R K G S P C(24) of human SRC-1. This immunizing peptide (Cat. # PEP-044) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | SRC1 |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | NCOA1 |
| Gene Accession No. | P70365, Q15788 |
| Gene Alias | ASV; bHLHe42; bHLHe74; class E basic helix-loop-helix protein 74; c-SRC; F-SRC-1; Hin-2 protein; KAT13A; mNRC-1; NCOA1; NCoA-1; Nuclear receptor coactivator 1; nuclear receptor coactivator protein 1; p60-Src; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; Src1; SRC-1; SRC-1 (-Q); SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1 |
| Gene Symbols | NCOA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
