Learn More
ABclonal Technology SREBF1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A26708100UL
Encompass_Preferred
This gene encodes a transcription factor that binds to the sterol regulatory element-1 (SRE1) which is a decamer flanking the low density lipoprotein receptor gene and some genes involved in sterol biosynthesis The protein is synthesized as a precursor that is attached to the nuclear membrane and endoplasmic reticulum Following cleavage the mature protein translocates to the nucleus and activates transcription by binding to the SRE1 Sterols inhibit the cleavage of the precursor and the mature nuclear form is rapidly catabolized thereby reducing transcription The protein is a member of the basic helix-loop-helix-leucine zipper (bHLH-Zip) transcription factor family Alternatively spliced transcript variants have been characterized for this gene
Catalog No. 50-000-11191