Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SREBP-2 Monoclonal Antibody (IgG-1C6), Active Motif™
SDP

Catalog No. 501992983
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2983 100 μg
50-199-2984 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2983 Supplier Active Motif Supplier No. 39941
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.

Specifications

Antigen SREBP-2
Applications Western Blot
Classification Monoclonal
Clone IgG-1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene SREBF2
Gene Accession No. Q12772
Gene Alias AI608257; BHLHD2; Class D basic helix-loop-helix protein 2; EMBL:AAI01903.1}; lop13; nuc; Processed sterol regulatory element-binding protein 2; srebf2; srebf2 {ECO:0000312; Srebf2_retired; SREBP2; SREBP-2; SREBP2gc; sterol regulatory element binding factor 2; sterol regulatory element binding protein 2; sterol regulatory element binding transcription factor 2; Sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; zgc:158371
Gene Symbols SREBF2
Host Species Mouse
Immunogen This SREBP-2 antibody was raised against a recombinant protein corresponding to amino acids 833-1141 of human SREBP-2.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6721
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.