Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SREBP1 Monoclonal Antibody (2A4)
GREENER_CHOICE

Catalog No. PIMA511685 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511685 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511685 Supplier Invitrogen™ Supplier No. MA511685
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-11685 targets SREBP-1 in WB applications and shows reactivity with Hamster and Human samples. The MA5-11685 immunogen is recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1.

Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SREBP1
Applications Western Blot
Classification Monoclonal
Clone 2A4
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene SREBF1
Gene Accession No. P36956, Q60416
Gene Alias ADD1; ADD-1; Adipocyte determination- and differentiation-dependent factor 1; bHLHd1; Class D basic helix-loop-helix protein 1; D630008H06; I79_010155; nt-SREBP1; Processed sterol regulatory element-binding protein 1; SREBF1; SREBP1; SREBP-1; SREBP1a; SREBP-1a; SREBP1c; SREBP-1c; sterol regulatory element binding protein 1; sterol regulatory element binding protein-1; sterol regulatory element binding transcription factor 1; sterol regulatory element binding-protein 1; sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1
Gene Symbols SREBF1
Host Species Mouse
Immunogen Recombinant protein encoding N-terminal amino acids 301-407 (the bHLH/leucine zipper domain) of human SREBP-1.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100689018, 6720
Target Species Hamster, Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.