Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SREBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524167
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524167 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524167 Supplier Invitrogen™ Supplier No. PA524167
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SREBP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SREBP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SREBF2
Gene Accession No. Q12772
Gene Alias AI608257; bHLHd2; Class D basic helix-loop-helix protein 2; EMBL:AAI01903.1}; lop13; nuc; Processed sterol regulatory element-binding protein 2; srebf2; srebf2 {ECO:0000312; Srebf2_retired; SREBP2; SREBP-2; SREBP2gc; sterol regulatory element binding factor 2; sterol regulatory element binding protein 2; sterol regulatory element binding transcription factor 2; sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; zgc:158371
Gene Symbols SREBF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 399-427 amino acids from the central region of human SREBF2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6721
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.