Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRGAP2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144229
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144229 Supplier Invitrogen™ Supplier No. PA5144229
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SRGAP2C Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Human-specific protein that acts as a key modifier of cortical connectivity in the human brain (PubMed:22559944, PubMed:27373832, PubMed:34707291). Acts by inhibiting the functions of ancestral paralog SRGAP2/SRGAP2A, a postsynaptic protein that regulates excitatory and inhibitory synapse maturation and density in cortical pyramidal neurons (PubMed:22559944, PubMed:27373832). SRGAP2C is unstable but is able to heterodimerize with SRGAP2/SRGAP2A, thereby reducing SRGAP2/SRGAP2A levels through proteasome-dependent degradation (PubMed:27373832, PubMed:28333212, PubMed:31822692). Inhibition of SRGAP2/SRGAP2A by SRGAP2C leads to an increase in synaptic density and protracted synaptic maturation of both excitatory and inhibitory synapses (PubMed:27373832, PubMed:34707291). Modifies cortical circuit connectivity by increasing the number of local and long-range cortical inputs received by layer 2/3 pyramidal neurons (PubMed:34707291). Also able to increase the probability of sensory-evoked responses by layer 2/3 pyramidal neurons (PubMed:34707291).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRGAP2C
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene SRGAP2C
Gene Accession No. P0DJJ0
Gene Alias SRGAP2P1
Gene Symbols SRGAP2C
Host Species Rabbit
Immunogen Recombinant Human SLIT-ROBO Rho GTPase-activating protein 2C protein (1-242AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653464
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.