Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRRD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551782
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551782 Supplier Invitrogen™ Supplier No. PA551782
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEKDLFISDF WSSALETINR CLTKHLEQLK APVGTLSDIF GNLHLDSLPE ESDVATDSIP REILVTGTCH LKCVCYGIGN FATCIVARNQ LTFLLLLLEK CQIPRSHCWV YDPLFSQLEI EVLNTLGVTV LSENEEGKRS I Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.

SRRD may be involved in a circadian clock input pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRRD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRRD
Gene Accession No. Q9UH36
Gene Alias HC/HCC; hepatocellular carcinoma complicating hemochromatosis; LOW QUALITY PROTEIN: SRR1-like protein; SRR1 domain containing; SRR1 domain-containing protein; SRR1L; SRR1-like protein; SRRD
Gene Symbols SRRD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRRD. Recombinant protein control fragment (Product #RP-91494).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 402055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.