Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRrp35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569114 Supplier Invitrogen™ Supplier No. PA569114
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 85%; Rabbit: 100%; Rat: 100%.

SRSF12 gene ontology annotations related to this gene include regulation of alternative mRNA splicing, via spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRrp35
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SRSF12
Gene Accession No. Q8WXF0
Gene Alias 35 kDa serine-arginine repressor; 35 kDa SR repressor protein; 35 kDa SR repressor protein-like protein; B830039L16; RGD1561162; RP11-63L7.3; serine and arginine rich splicing factor 12; serine/arginine-rich splicing factor 12; serine/arginine-rich splicing factor 13B; serine-arginine repressor protein; serine-arginine repressor protein (35 kDa); serine-arginine repressor protein 35; Sfrs13b; SFRS19; splicing factor, arginine/serine-rich 13B; Splicing factor, arginine/serine-rich 19; SR splicing factor 12; Srrp; SRRP35; Srsf12; Srsf13b
Gene Symbols SRSF12
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SRrp35.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135295
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.