Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SRSF11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101910 Supplier Invitrogen™ Supplier No. PA5101910
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFRS11.

Pre-mRNA splicing enhancer elements are short RNA sequences capable of activating weak splice sites in nearby introns that are required for accurate splice site recognition and the control of alternative splicing. Splicing enhancer elements contain specific binding sites for serine/arginine (SR)-rich splicing factors, which include SC35, 9G8, SRp20 and SF2/ASF. The family of SR factors all contain one or more RNA recognition motifs (RRM) and a SR-rich domain. The SR factor family is not only essential for constitutive splicing, but also regulate splicing in a concentration-dependent manner by influencing the selection of alternative splice sites. SFRS11 (splicing factor, arginine/serine-rich 11), also known as arginine-rich 54 kDa nuclear protein (p54) or NET2, is a 484 amino acid nuclear protein that colocalizes with spliceosome components and belongs to the splicing factor SR family. SFRS11 may function in pre-mRNA splicing and contains one RRM (RNA recognition motif) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SRSF11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SRSF11
Gene Accession No. Q05519
Gene Alias 0610009J05Rik; 2610019N13Rik; Arginine-rich 54 kDa nuclear protein; BF642805; dJ677H15.2; NET2; p54; serine and arginine rich splicing factor 11; serine/arginine-rich splicing factor 11; SFR11; Sfrs11; splicing factor p54; splicing factor, arginine/serine-rich 11; SR splicing factor 11; Srsf11
Gene Symbols SRSF11
Host Species Rabbit
Immunogen A synthesized peptide derived from human SRSF11(Accession Q05519), corresponding to amino acid residues E441-D484.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69207, 9295
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.