Learn More
Invitrogen™ SRSF9 Monoclonal Antibody (OTI5G7)

Description
SRSF9 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SRSF9 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5G7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SRSF9 |
| Gene Accession No. | Q13242 |
| Gene Alias | 25kDa; 2610029M16Rik; hypothetical protein zgc:77449; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 9; pre-mRNA-splicing factor SRp30C; serine and arginine rich splicing factor 9; serine/arginine-rich splicing factor 9; Sfrs9; splicing factor, arginine/serine rich 9; splicing factor, arginine/serine-rich 9; SR splicing factor 9; SRP30C; Srsf9; zgc:77449 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.