Learn More
Selleck Chemical LLC SRT1720 HCl 5mg 1001645-58-4
SDP
Supplier:  Selleck Chemical LLC S11295MG
Encompass_Preferred
SRT1720 HCl is a selective SIRT1 activator with EC50 of 0.16 ?M in a cell-free assay, but is >230-fold less potent for SIRT2 and SIRT3. SRT1720 induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0248054
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.