Learn More
Electron Microscopy Sciences Sure Shot Atomizer Model 8500PL Stainless Steel, Plastic Top 16 OZ
SDP
Supplier:  Electron Microscopy Sciences 6418610
Encompass_Preferred
Anything that the aerosol can do, ATOMIZER can do safer. Refillable and Rechargeable. Eliminates aerosol disposal problems. All Model B sprayers come standard with Viton seals, these seals are compatible with most products. IMPORTANT: Sure Shot Sprayers are not recommended for spraying any acids. Sure Shot Sprayers will handle most chemicals, but are not designed to be used with products that are intended to etch or attack metal surfaces. Stainless Steel with plastic top. Comes with extra valve and 2 fine, 2 regular, medium, 2 coarse and 1 pin stream nozzles.
Catalog No. 50-288-40